4-Nitrophenethyl bromide 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1-(2-溴乙基)-4-硝基苯, 2-(4-硝基苯基)乙基溴
151620050
151620250
ACRO151620050EA
0
CNY
ACRO151620050
ACRO151620250
4-Nitrophenethyl bromide 98%
4-硝基苯乙基溴
分子式:
C₈H₈BrNO₂ 分子量: 230.06 g/mol 溶点: 67…70 °C 存储条件: Ambient |
MDL编号:
MFCD00007386 CAS号: 5339-26-4 EINECS: 226-271-7 |
产品技术规格
Appearance | Yellow Adhering crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 67 to 70 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...