N-甲酰吗啉 99+%

供应商: Thermo Fisher Scientific

Synonyms: 4-吗啉甲醛

133800050 133801000 133805000
ACRO133800050EA 0 CNY
ACRO133800050 ACRO133801000 ACRO133805000
N-甲酰吗啉 99+%
N-甲酰吗啉
分子式: C₅H₉NO₂
溶点: 23 °C
密度: 1,14 g/cm³
闪点: 113 °C
存储条件: Ambient
MDL编号: MFCD00006170
CAS号: 4394-85-8
EINECS: 224-518-3

现在订购

产品技术规格

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4855 to 1.4875 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR