4-氨基吗啉 95%

供应商: Thermo Fisher Scientific
Warning

Synonyms: N-Aminomorpholine

104110250 104110050
ACRO104110250EA 0 CNY
ACRO104110250 ACRO104110050
4-氨基吗啉 95%
4-氨基吗啉
分子式: C₄H₁₀N₂O
分子量: 102.14 g/mol
密度: 1.05 g/cm³
闪点: 58 °C
存储条件: Ambient
MDL编号: MFCD00006174
CAS号: 4319-49-7
EINECS: 224-347-4
UN号: 1993
ADR: 3 III

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4800 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR