2-溴-1,4-二甲苯 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-溴-对二甲苯
173420250
173421000
ACRO173420250EA
0
CNY
ACRO173420250
ACRO173421000
2-溴-1,4-二甲苯 97%
2-溴-1,4-二甲苯
分子式:
C₈H₉Br 分子量: 185.06 g/mol 沸点: 200 溶点: 9…10 °C 密度: 1.34 g/cm³ 闪点: 79 °C |
MDL编号:
MFCD00000074 CAS号: 553-94-6 EINECS: 209-055-7 |
产品技术规格
Appearance | Clear colorless to slightly colored Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5495 to 1.5515 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...