α,α-二氯二苯甲烷 97%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
二氯二苯甲酮
113330500
113332500
ACRO113330500EA
0
CNY
ACRO113330500
ACRO113332500
α,α-二氯二苯甲烷 97%
α,α-二氯二苯甲烷
分子式:
(C₆H₅)₂CCl₂ 分子量: 237.13 g/mol 沸点: 305 °C (760 mmHg) 溶点: 148 密度: 1.23 g/cm³ 闪点: >112 °C 存储条件: Ambient |
MDL编号:
MFCD00000811 CAS号: 2051-90-3 EINECS: 218-134-5 UN号: 3265 ADR: 8 II |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6040 to 1.6060 (20°C, 589 nm) |
GC | ≥96.0 % |
Uniformity coefficient | ≤1.8 |
Moisture holding capacity | 53 to 58 % (H+) |
Total volume capacity | ≥1.8 eq/l (H+) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...