Octadecylbenzene 98%
供应商: Thermo Fisher Scientific
Synonyms:
1-Phenyloctadecane
124700010
124700100
ACRO124700010EA
0
CNY
ACRO124700010
ACRO124700100
Octadecylbenzene 98%
Octadecylbenzene
分子式:
C₂₄H₄₂ 分子量: 330.6 g/mol 沸点: 249 °C (15 mmHg) 溶点: 32…36 °C 闪点: >112 °C 存储条件: Ambient |
MDL编号:
MFCD00048500 CAS号: 4445-07-2 EINECS: 224-684-7 |
产品技术规格
Appearance | White to light yellow Low melting solid |
Infrared spectrum | Conforms |
Melting point | 32 to 36 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...