4-碘三氟甲苯 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-碘-α,α,α-三氟甲苯
306180050
306180010
ACRO306180050EA
0
CNY
ACRO306180050
ACRO306180010
4-碘三氟甲苯 97%
4-碘三氟甲苯
分子式:
C₇H₄F₃I 沸点: 76…79 °C (15 mmHg) 溶点: 17 °C 密度: 1,851 g/cm³ 闪点: 68 °C |
MDL编号:
MFCD00039398 CAS号: 455-13-0 EINECS: 207-234-4 |
产品技术规格
Appearance | Clear colorless to yellow to red Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5160 to 1.5200 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...