对氟甲苯 97%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1-Fluoro-4-methylbenzene
119670100
119671000
119672500
ACRO119670100EA
0
CNY
ACRO119670100
ACRO119671000
ACRO119672500
对氟甲苯 97%
对氟甲苯
分子式:
C₇H₇F 分子量: 110.13 g/mol 沸点: 116 溶点: –56 °C 密度: 1 g/cm³ 闪点: 10 °C 存储条件: Ambient |
MDL编号:
MFCD00000358 CAS号: 352-32-9 EINECS: 206-520-6 UN号: 2388 ADR: 3 II Merck Index: 13,04207 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4674 to 1.4694 (20°C, 589 nm) |
Water | ≤0.05 % (K.F.) |
GC | ≥96.0 % |
Specific optical rotation | +9.5° to +11.5° (20°C, 589 nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...