2-氟甲苯 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 1-Fluoro-2-methylbenzene, o-Fluorotoluene

119661000 119665000
ACRO119661000EA 0 CNY
ACRO119661000 ACRO119665000
2-氟甲苯 99%
2-氟甲苯
分子式: C₇H₇F
分子量: 110.13 g/mol
沸点: 113…114 °C (760 mmHg)
溶点: –62 °C
密度: 1 g/cm³
闪点: 8 °C
存储条件: Ambient
MDL编号: MFCD00000322
CAS号: 95-52-3
EINECS: 202-428-5
UN号: 2388
ADR: 3 II
Merck Index: 13,04207

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4750 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure
Specific optical rotation -9.5° to -11.5° (20°C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR