4-硝基苄氯 99%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
α-氯-4-硝基甲苯
161090100
161091000
ACRO161090100EA
0
CNY
ACRO161090100
ACRO161091000
4-硝基苄氯 99%
4-硝基苄氯
分子式:
C₇H₆ClNO₂ 分子量: 171.58 g/mol 沸点: 112 °C (.6 mmHg) 溶点: 71…74 °C 密度: 1.5647 存储条件: Ambient |
MDL编号:
MFCD00007374 CAS号: 100-14-1 EINECS: 202-822-7 UN号: 3261 ADR: 8 II |
产品技术规格
Appearance | Light yellow Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 71 to 74 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...