4-氯氯苄 99+%

供应商: Thermo Fisher Scientific
Warning

Synonyms: α,4-二氯甲苯

150240050 150242500
ACRO150240050EA 0 CNY
ACRO150240050 ACRO150242500
4-氯氯苄 99+%
4-氯氯苄
分子式: C₇H₆Cl₂
分子量: 161.03 g/mol
沸点: 221 °C (760 mmHg)
溶点: 27…28 °C
闪点: 107 °C
存储条件: Ambient
MDL编号: MFCD00000914
CAS号: 104-83-6
EINECS: 203-242-7
UN号: 3427
ADR: 6.1 III

现在订购

产品技术规格

Appearance Colorless to white Low melting crystalline solid
Infrared spectrum Conforms
Melting point ≥28 °C
GC ≤0.5 % p-Chlorobenzal chloride


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR