4-甲基水杨酸 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 间甲基水杨酸, 2-羟基-4-甲基苯甲酸

223540250 223541000
ACRO223540250EA 0 CNY
ACRO223540250 ACRO223541000
4-甲基水杨酸 98%
4-甲基水杨酸
分子式: C₈H₈O₃
分子量: 152.15 g/mol
溶点: 175…179 °C
MDL编号: MFCD00002454
CAS号: 50-85-1
EINECS: 200-068-3

现在订购

产品技术规格

Appearance White to grey-beige Powder
Infrared spectrum Conforms
Melting point 171 to 179 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR