3-甲基水杨酸 99%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-羟基-3-甲基苯甲酸, 邻甲基水杨酸
173340050
173345000
ACRO173340050EA
0
CNY
ACRO173340050
ACRO173345000
3-甲基水杨酸 99%
3-甲基水杨酸
分子式:
C₈H₈O₃ 分子量: 152.15 g/mol 沸点: 293 溶点: 164…167 °C 存储条件: Ambient |
MDL编号:
MFCD00002448 CAS号: 83-40-9 EINECS: 201-473-8 Merck Index: 13,02609 |
产品技术规格
Appearance | White to off-white to light pink Powder |
Infrared spectrum | Conforms |
Melting point | 163 to 167 °C |
Water | ≤0.3 % (K.F.) |
Titration with NaOH | 98.5 to 101.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...