2-氨基-5-甲基苯甲酸 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 5-甲基邻氨基苯甲酸

160200010 160200050
ACRO160200010EA 0 CNY
ACRO160200010 ACRO160200050
2-氨基-5-甲基苯甲酸 97%
2-氨基-5-甲基苯甲酸
分子式: C₈H₉NO₂
分子量: 151.16 g/mol
沸点: 317
溶点: 174…177 °C (dec.)
存储条件: Ambient
MDL编号: MFCD00007909
CAS号: 2941-78-8
EINECS: 220-932-3

现在订购

产品技术规格

Appearance Off-white to beige to light brown Powder
Infrared spectrum Conforms
Melting point 173 to 178 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR