4-(4-羟基苯基)-2-丁酮 99+%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 覆盆子酮

176470500 176472500
ACRO176470500EA 0 CNY
ACRO176470500 ACRO176472500
4-(4-羟基苯基)-2-丁酮 99+%
4-(4-羟基苯基)-2-丁酮
分子式: C₁₀H₁₂O₂
分子量: 164.2 g/mol
沸点: 292
溶点: 82…84 °C
存储条件: Ambient
MDL编号: MFCD00002394
CAS号: 5471-51-2
EINECS: 226-806-4

现在订购

产品技术规格

Appearance White to light yellow Powder or needles
Infrared spectrum Conforms
Melting point 81 to 85 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR