苯戊酮 98%
供应商: Thermo Fisher Scientific
Synonyms:
丁基苯酮
140800250
140801000
140800050
ACRO140800250EA
0
CNY
ACRO140800250
ACRO140801000
ACRO140800050
苯戊酮 98%
苯戊酮
分子式:
C₆H₅C(O)(CH₂)₃CH₃ 分子量: 162.23 g/mol 沸点: 105…107 °C (5 mmHg) 溶点: –9 °C 密度: 0.98 g/cm³ 闪点: 102 °C 存储条件: Ambient |
MDL编号:
MFCD00009480 CAS号: 1009-14-9 EINECS: 213-767-3 |
|
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5130 to 1.5170 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...