4-硝基苄溴 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: α-溴-4-硝基甲苯

128530250 128531000
ACRO128530250EA 0 CNY
ACRO128530250 ACRO128531000
4-硝基苄溴 99%
4-硝基苄溴
分子式: C₇H₆BrNO₂
分子量: 216.03 g/mol
沸点: 304
溶点: 98…100 °C
闪点: >100 °C
存储条件: Ambient
MDL编号: MFCD00007373
CAS号: 100-11-8
EINECS: 202-820-6
UN号: 3261
ADR: 8 II

现在订购

产品技术规格

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 100 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR