2-硝基苄溴 97%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
α-溴-2-硝基甲苯
148310050
148310250
ACRO148310050EA
0
CNY
ACRO148310050
ACRO148310250
2-硝基苄溴 97%
2-硝基苄溴
分子式:
C₇H₆BrNO₂ 分子量: 216.03 g/mol 溶点: 44…48 °C 闪点: >110 °C 存储条件: Refrigerator |
MDL编号:
MFCD00007184 CAS号: 3958-60-9 EINECS: 223-558-9 UN号: 3261 ADR: 8 II |
产品技术规格
Appearance | Yellow to light brown Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 44 to 48 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...