邻氟溴苄 98%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-氟溴苄
197960050
197960250
ACRO197960050EA
0
CNY
ACRO197960050
ACRO197960250
邻氟溴苄 98%
邻氟溴苄
分子式:
C₇H₆BrF 分子量: 189.03 g/mol 沸点: 84…85 °C (15 mmHg) 密度: 1.567 g/cm³ 闪点: 82 °C 存储条件: Ambient |
MDL编号:
MFCD00000324 CAS号: 446-48-0 EINECS: 207-169-1 UN号: 3265 ADR: 8 II |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5495 to 1.5515 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...