4-吡啶硼酸频哪醇酯 97%

供应商: Thermo Fisher Scientific
Warning

367510010 367510050
ACRO367510010EA 0 CNY
ACRO367510010 ACRO367510050
4-吡啶硼酸频哪醇酯 97%
4-吡啶硼酸频哪醇酯
分子式: C₁₁H₁₆BNO₂
溶点: 153…155 °C
存储条件: Refrigerator
MDL编号: MFCD01319051
CAS号: 181219-01-2

现在订购

产品技术规格

Appearance White to off-white Powder
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR