1-溴-3-苯基丙烷 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: (3-溴丙基)苯, Hydrocinnamyl bromide, 3-苯丙基溴

107290050 107291000 107295000
ACRO107290050EA 0 CNY
ACRO107290050 ACRO107291000 ACRO107295000
1-溴-3-苯基丙烷 98%
1-溴-3-苯基丙烷
分子式: BrH₂CCH₂CH₂(C₆H₅)
分子量: 199.09 g/mol
沸点: 219…220 °C (760 mmHg)
密度: 1.31 g/cm³
闪点: 101 °C
存储条件: Ambient
MDL编号: MFCD00000257
CAS号: 637-59-2
EINECS: 211-294-7

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR