2-Naphthalenethiol 98+%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
硫代-2-萘酚 (β), 2-萘硫酚
334520050
334520250
ACRO334520050EA
0
CNY
ACRO334520050
ACRO334520250
2-Naphthalenethiol 98+%
2-萘硫醇
分子式:
C₁₀H₈S 分子量: 160.24 g/mol 沸点: 286 °C 溶点: 79…83 °C 存储条件: Ambient |
MDL编号:
MFCD00004086 CAS号: 91-60-1 EINECS: 202-082-5 |
产品技术规格
Appearance | White to light beige to light grey Powder or crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 79 to 83 °C |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...