(R)-(+)-1-(1-萘基)乙胺 99+%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(R)-(+)-α-甲基-1-萘甲胺
217800010
217800050
ACRO217800010EA
0
CNY
ACRO217800010
ACRO217800050
(R)-(+)-1-(1-萘基)乙胺 99+%
(R)-(+)-1-(1-萘基)乙胺
分子式:
C₁₂H₁₃N 分子量: 171.24 g/mol 沸点: 153 °C (11 mmHg) 闪点: >110 °C 存储条件: Ambient |
MDL编号:
MFCD00064114 CAS号: 3886-70-2 EINECS: 223-425-5 |
产品技术规格
Appearance | Clear colorless to yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.6215 to 1.6245 (20°C, 589 nm) |
GC | ≥99.0 % |
Specific optical rotation | +51° to +59° (20°C, 589 nm) (c=2, ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...