(S)-2-氯-3-甲基丁酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: (S)-α-氯代异戊酸

C1365-1G C1365-5G
TCIAC1365-1GEA 0 CNY
TCIAC1365-1G TCIAC1365-5G
(S)-2-氯-3-甲基丁酸 ≥98.0% (by titrimetric analysis)
(S)-2-氯-3-甲基丁酸
分子式: C₅H₉ClO₂
分子量: 136.58 g/mol
沸点: 109°C
MDL编号: MFCD00067109
CAS号: 26782-74-1
UN号: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a68ce75099536601bf36d365b916e08e3d021e6&vid=5a68ce75099536601bf36d365b916e08e3d021e6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %
Optical purity (GC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -1.0 to -2.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR