N-哌嗪甲酸乙酯 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 1-乙氧基羰基哌嗪, 1-哌嗪羧酸乙酯

118870250 118871000
ACRO118870250EA 0 CNY
ACRO118870250 ACRO118871000
N-哌嗪甲酸乙酯 99%
N-哌嗪甲酸乙酯
分子式: C₇H₁₄N₂O₂
分子量: 158.2 g/mol
沸点: 273 °C (760 mmHg)
密度: 1.08 g/cm³
闪点: >110 °C
存储条件: Ambient
MDL编号: MFCD00005964
CAS号: 120-43-4
EINECS: 204-395-2

现在订购

产品技术规格

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4790 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR