5-Chlorotryptamine hydrochloride ≥98.0% (以总氮计)
供应商: TCI
|
Warning
|
C1145-25G
C1145-5G
TCIAC1145-25GEA
0
CNY
TCIAC1145-25G
TCIAC1145-5G
5-Chlorotryptamine hydrochloride ≥98.0% (以总氮计)
5-Chlorotryptamine hydrochloride
分子式:
C₁₀H₁₂Cl₂N₂ |
MDL编号:
MFCD00051990 CAS号: 942-26-7 EINECS: 213-387-8 |
|
产品技术规格
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...