三甲基碘化亚砜 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
Trimethyloxosulphonium iodide
T1065-25G
T1065-500G
TCIAT1065-25GEA
0
CNY
TCIAT1065-25G
TCIAT1065-500G
三甲基碘化亚砜 ≥98.0% (by titrimetric analysis)
三甲基碘化亚砜
分子式:
C₃H₉IOS 分子量: 220.07 g/mol |
MDL编号:
MFCD00011899 CAS号: 1774-47-6 EINECS: 217-204-2 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...