Dressing jars, with lid
供应商: BOCHEM
8581 8584 8585 8587 8588 8693
BOCH8581EA 0 CNY
BOCH8581 BOCH8584 BOCH8585 BOCH8587 BOCH8588 BOCH8693
Dressing jars, with lid
通用罐
Jar made of stainless steel, with knob lid.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean