3,4-二甲氧基苯胺 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-Aminoveratrole, 1,2-二甲氧基-4-氨基苯
104720100
104720250
ACRO104720100EA
0
CNY
ACRO104720100
ACRO104720250
3,4-二甲氧基苯胺 98%
3,4-二甲氧基苯胺
分子式:
C₈H₁₁NO₂ 分子量: 153.18 g/mol 沸点: 174…176 °C (22 mmHg) 溶点: 86…88 °C 闪点: >110 °C 存储条件: Refrigerator |
MDL编号:
MFCD00008394 CAS号: 6315-89-5 EINECS: 228-647-6 UN号: 2811 ADR: 6.1 III |
产品技术规格
Appearance | Brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 86 to 88 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...