对碘苯甲醚 98%

供应商: Thermo Fisher Scientific

Synonyms: 对甲氧基碘代苯, 4-碘苯甲醚

122320250 122321000
ACRO122320250EA 0 CNY
ACRO122320250 ACRO122321000
对碘苯甲醚 98%
对碘苯甲醚
分子式: C₇H₇IO
沸点: 237 °C (726 mmHg)
溶点: 46…51 °C
密度: 1 g/cm³ (20 °C)
闪点: >110 °C
存储条件: Ambient
MDL编号: MFCD00001056
CAS号: 696-62-8
EINECS: 211-798-7

现在订购

产品技术规格

Appearance Off-white to brown Crystalline powder and/or chunks (May darken on storage)
Infrared spectrum Conforms
Melting point 46 to 51 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR