3-碘苯甲醚 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
3-iodoanisole
222020050
222020250
ACRO222020050EA
0
CNY
ACRO222020050
ACRO222020250
3-碘苯甲醚 97%
3-碘苯甲醚
分子式:
C₇H₇IO 沸点: 244…245 °C (760 mmHg) 密度: 1,96 g/cm³ 闪点: >110 °C 存储条件: Ambient |
MDL编号:
MFCD00001048 CAS号: 766-85-8 EINECS: 212-173-1 |
产品技术规格
Appearance | Clear yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.6130 to 1.6150 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...