间碘苯甲酸 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: m-Iodobenzoic acid

160480100 160480500
ACRO160480100EA 0 CNY
ACRO160480100 ACRO160480500
间碘苯甲酸 98%
间碘苯甲酸
分子式: C₇H₅IO₂
沸点: 337 °C (1013 hPa)
溶点: 185…189 °C
密度: 2,171 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00002496
CAS号: 618-51-9
EINECS: 210-555-2

现在订购

产品技术规格

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 185 to 189 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR