五氟苯酚 99+%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 2,3,4,5,6-五氟苯酚

147130050 147130100
ACRO147130050EA 0 CNY
ACRO147130050 ACRO147130100
五氟苯酚 99+%
五氟苯酚
分子式: C₆HF₅O
沸点: 143 °C (760 mmHg)
溶点: 34…36 °C
密度: 1,75 g/cm³ (20 °C)
闪点: 72 °C
存储条件: Ambient
MDL编号: MFCD00002156
CAS号: 771-61-9
EINECS: 212-235-8
UN号: 2923
ADR: 8,III

现在订购

产品技术规格

Appearance White to off-white Low melting crystalline solid
Infrared spectrum Conforms
GC ≥99.0 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR