对氟硝基苯 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-Fluoronitrobenzene, 1-Fluoro-4-nitrobenzene
119491000
119495000
ACRO119491000EA
0
CNY
ACRO119491000
ACRO119495000
对氟硝基苯 99%
对氟硝基苯
分子式:
C₆H₄FNO₂ 分子量: 141.1 g/mol 沸点: 205 溶点: 27 °C 密度: 1.33 g/cm³ 闪点: 83 存储条件: Ambient |
MDL编号:
MFCD00007282 CAS号: 350-46-9 EINECS: 206-502-8 UN号: 2811 ADR: 6.1 III |
产品技术规格
Appearance | Clear yellow to dark brown Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.5302 to 1.5322 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...