2,4-二氟苯胺 99%
供应商: Thermo Fisher Scientific
|
Danger
|
114590100
114590250
ACRO114590100EA
0
CNY
ACRO114590100
ACRO114590250
2,4-二氟苯胺 99%
2,4-二氟苯胺
分子式:
C₆H₅F₂N 沸点: 170 °C (753 mmHg) 密度: 1,268 g/cm³ 闪点: 62 °C 存储条件: Refrigerator |
MDL编号:
MFCD00007648 CAS号: 367-25-9 EINECS: 206-687-5 UN号: 2941 ADR: 6.1,III Merck Index: 13,03171 |
产品技术规格
Appearance | Clear colorless to yellow to brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5055 to 1.5085 (20°C, 589 nm) |
GC | ≥98.5 % |
Stabilizer | ≤1.1 % Triethylamine |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...