间氟碘苯 99% stabilised

供应商: Thermo Fisher Scientific
Warning

Synonyms: 3-Fluoroiodobenzene

204480050 204480250
ACRO204480050EA 0 CNY
ACRO204480050 ACRO204480250
间氟碘苯 99% stabilised
间氟碘苯
分子式: C₆H₄FI
分子量: 222 g/mol
沸点: 77…78 °C (19 mmHg)
溶点: –42 °C
密度: 1.89 g/cm³
闪点: 67 °C
MDL编号: MFCD00001044
CAS号: 1121-86-4
EINECS: 214-339-9

现在订购

产品技术规格

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5830 to 1.5860 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Copper


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR