Specimen containers, with lid, SecurTainer™ I
供应商: SIMPORT SCIENTIFIC
C575-40MA C575-120MA C575-20MA C575-60MA C575-90MA
SIMPC575-40MAEA 0 CNY
SIMPC575-40MA SIMPC575-120MA SIMPC575-20MA SIMPC575-60MA SIMPC575-90MA
Specimen containers, with lid, SecurTainer™ I
样品容器 通用容器
Tamper evident specimen containers, with closure, uniquely stackable for collection, transport and storage of specimens.
  • Shatter resistant, eliminating most problems of leakage and evaporation
  • Safe storage and almost transparent to allow specimens to be viewed without opening
  • Manufactured from virgin PP with a unique integrated leak-resistant seal closure, PE
  • All containers are 95 kpa compliant
  • Functional temperature range from –90 to +100 °C
The magenta lids are ribbed for easy opening when hands are wet or gloved while the jars are stackable.
Packaging: Containers and caps packaged separately in bags of 100