1-氯-4-氟苯 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: Chloro-1-fluoro-4-benzène, 4-Chlorofluorobenzene

208200250 208200050
ACRO208200250EA 0 CNY
ACRO208200250 ACRO208200050
1-氯-4-氟苯 98%
1-氯-4-氟苯
分子式: C₆H₄ClF
分子量: 130.55 g/mol
沸点: 129
溶点: –21.5 °C
密度: 1.226 g/cm³
闪点: 29 °C
MDL编号: MFCD00000603
CAS号: 352-33-0
EINECS: 206-521-1
UN号: 1993

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4960 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR