Ultra-microbalances and Microbalances, Excellence XP Series
供应商: METTLER TOLEDO
11122415 11122430 11106048 11106051 11106054 11106057 11122400
METB11122415EA 0 CNY
METB11122415 METB11122430 METB11106048 METB11106051 METB11106054 METB11106057 METB11122400
Ultra-microbalances and Microbalances, Excellence XP Series
天平
A range of high performance microbalances with the XP interface that delivers levels of control and safety that are required for regulated environments.
  • proFACT fully automatic time and/or temperature controlled internal adjustment
  • MinWeigh warns if minimum weight requirement is violated
  • RS232C with two auxiliary interfaces for connecting a keyboard or additional sensor
  • Slot for second optional interface including USB, Bluetooth and Ethernet
Ultra-microbalances: Seven figure (0,1 µg) readability offers the user the accuracy and tolerances to measure samples below 1 mg and still comply with international regulatory standards.
*Reliability of result with precious samples
*Simple to use, with two SmartSens infrared sensors for hands free operation
Microbalance with round weigh cell has similar features and design to the ultra microbalances
Microbalances with automatic, square, dual draught shields and SmartGrid are a range of balances for microdosing directly into larger containers.
*Fast stabilisation times 2,5 to 3,5 seconds
*Automatic, individually configurable, dual draught shields reduce air turbulence
*SmartGrid micro for more secure weighing and easy cleaning
*ErgoClips micro allow secure fastening of small and odd shaped containers
*MinWeigh door micro allows dosing into tare container without opening the draught shield
订购须知 For details of the full range of accessories, including options for filter weighing and antistatic options, please contact Avantor.
Caution: * Measured at low load conditions (0,2 g)
** Measured at low load conditions (1 g)