3-氯苯甲醛 99%
供应商: Thermo Fisher Scientific
|
Warning
|
108610250
108611000
ACRO108610250EA
0
CNY
ACRO108610250
ACRO108611000
3-氯苯甲醛 99%
3-氯苯甲醛
分子式:
C₇H₅ClO 分子量: 140.57 g/mol 沸点: 213…215 °C (760 mmHg) 溶点: 17…18 °C 密度: 1.241 g/cm³ 闪点: 88 °C 存储条件: Ambient |
MDL编号:
MFCD00003350 CAS号: 587-04-2 EINECS: 209-596-9 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5630 to 1.5650 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...