3,4-二羟基苯甲酸 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
原儿茶酸
114890250
114891000
ACRO114890250EA
0
CNY
ACRO114890250
ACRO114891000
3,4-二羟基苯甲酸 97%
3,4-二羟基苯甲酸
分子式:
C₇H₆O₄ 分子量: 154.12 g/mol 溶点: 198…202 °C 存储条件: Ambient |
MDL编号:
MFCD00002509 CAS号: 99-50-3 EINECS: 202-760-0 Merck Index: 12,08080 |
产品技术规格
Appearance | Beige to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 198 to 202 °C |
HPLC | ≥96.0 % |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...