2,5-二羟基苯甲酸 99%
供应商: Thermo Fisher Scientific
|
Warning
|
165200050
165200500
ACRO165200050EA
0
CNY
ACRO165200050
ACRO165200500
2,5-二羟基苯甲酸 99%
2,5-二羟基苯甲酸
分子式:
C₇H₆O₄ 沸点: 339 °C (1013 hPa) 溶点: 204…207 °C 密度: 1,55 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00002460 CAS号: 490-79-9 EINECS: 207-718-5 Merck Index: 13,04409 |
产品技术规格
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 204 to 207 °C |
HPLC | ≥98.5 % |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...