2,3-二羟基苯甲酸 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: Pyrocatechuic acid

155170050 155170250
ACRO155170050EA 0 CNY
ACRO155170050 ACRO155170250
2,3-二羟基苯甲酸 99%
2,3-二羟基苯甲酸
分子式: C₇H₆O₄
分子量: 154.12 g/mol
沸点: 339
溶点: 204…208 °C
存储条件: Ambient
MDL编号: MFCD00002446
CAS号: 303-38-8
EINECS: 206-139-5

现在订购

产品技术规格

Appearance Beige to pink to grey to brown Crystalline powder
Infrared spectrum Conforms
Melting point 204 to 208 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR