Metal hub needles
供应商: HAMILTON BONADUZ
90018 90019 90020 90021 90022 90023 90024 90025 90026 90027 90031 90033 90038 90039 91038 91039 91022 7751-12 7748-09 91033 7748-03 7748-08 91023 7729-07 7747-01 7748-02 7748-12 7748-14 7751-07 7751-17 91025 7748-17 7748-18 7748-06 7729-10 91024 91014 7729-03 7748-07 91026 91017 91018 91020 91021 7729-02 7729-04 7729-05 7729-06 7729-08 7729-09 7729-01 7748-16 7749-05 7748-11 7751-14 7749-02 7751-08 7751-09 7751-11 7751-13 7751-15 7751-16 7751-18 7751-19 7748-10 7749-01 7749-03 7749-04 7748-15 7748-05 7751-01 7747-02 7751-02 7751-03 7751-04 7751-05 7751-06 7751-10 7751-20 7751-21 7748-01 7748-20 91019
HAMI90018EA 0 CNY
HAMI90018 HAMI90019 HAMI90020 HAMI90021 HAMI90022 HAMI90023 HAMI90024 HAMI90025 HAMI90026 HAMI90027 HAMI90031 HAMI90033 HAMI90038 HAMI90039 HAMI91038 HAMI91039 HAMI91022 HAMI7751-12 HAMI7748-09 HAMI91033 HAMI7748-03 HAMI7748-08 HAMI91023 HAMI7729-07 HAMI7747-01 HAMI7748-02 HAMI7748-12 HAMI7748-14 HAMI7751-07 HAMI7751-17 HAMI91025 HAMI7748-17 HAMI7748-18 HAMI7748-06 HAMI7729-10 HAMI91024 HAMI91014 HAMI7729-03 HAMI7748-07 HAMI91026 HAMI91017 HAMI91018 HAMI91020 HAMI91021 HAMI7729-02 HAMI7729-04 HAMI7729-05 HAMI7729-06 HAMI7729-08 HAMI7729-09 HAMI7729-01 HAMI7748-16 HAMI7749-05 HAMI7748-11 HAMI7751-14 HAMI7749-02 HAMI7751-08 HAMI7751-09 HAMI7751-11 HAMI7751-13 HAMI7751-15 HAMI7751-16 HAMI7751-18 HAMI7751-19 HAMI7748-10 HAMI7749-01 HAMI7749-03 HAMI7749-04 HAMI7748-15 HAMI7748-05 HAMI7751-01 HAMI7747-02 HAMI7751-02 HAMI7751-03 HAMI7751-04 HAMI7751-05 HAMI7751-06 HAMI7751-10 HAMI7751-20 HAMI7751-21 HAMI7748-01 HAMI7748-20 HAMI91019
Metal hub needles
注射器针头
Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors. The needles are 304 stainless steel. Custom needle lengths and point styles are available.