2-羟基-5-硝基苯甲酸 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
5-硝基水杨酸
300480050
300480250
300482500
ACRO300480050EA
0
CNY
ACRO300480050
ACRO300480250
ACRO300482500
2-羟基-5-硝基苯甲酸 99%
2-羟基-5-硝基苯甲酸
分子式:
C₇H₅NO₅ 分子量: 183.12 g/mol 溶点: 228…231 °C 存储条件: Ambient |
MDL编号:
MFCD00007338 CAS号: 96-97-9 EINECS: 202-548-8 Merck Index: 12,06730 |
产品技术规格
Appearance | Off-white to yellow to beige to orange to tan Powder |
Infrared spectrum | Conforms |
Melting point | 228 to 233 °C |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...