3,4-二氨基苯甲酸 97%
供应商: Thermo Fisher Scientific
|
Warning
|
112100250
112101000
ACRO112100250EA
0
CNY
ACRO112100250
ACRO112101000
3,4-二氨基苯甲酸 97%
3,4-二氨基苯甲酸
分子式:
C₇H₈N₂O₂ 沸点: 354 °C (1013 hPa) 溶点: 215 °C (dec.) 密度: 1,544 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00007726 CAS号: 619-05-6 EINECS: 210-577-2 |
产品技术规格
Appearance | Brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with NaOH | 96.0 to 104.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...