2,4,5-三羟基苯甲醛 99%
供应商: Thermo Fisher Scientific
|
Warning
|
345940010
345940050
ACRO345940010EA
0
CNY
ACRO345940010
ACRO345940050
2,4,5-三羟基苯甲醛 99%
2,4,5-三羟基苯甲醛
分子式:
C₇H₆O₄ 分子量: 154.12 g/mol 溶点: 226…227 °C |
MDL编号:
MFCD00016592 CAS号: 35094-87-2 |
产品技术规格
Appearance | Yellow to brown to khaki Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...