2-羟基-4-甲氧基苯甲醛 99%
供应商: Thermo Fisher Scientific
|
Warning
|
171080050
171080250
ACRO171080050EA
0
CNY
ACRO171080050
ACRO171080250
2-羟基-4-甲氧基苯甲醛 99%
2-羟基-4-甲氧基苯甲醛
分子式:
C₈H₈O₃ 沸点: 124 °C (12 mmHg) 溶点: 40…43 °C 密度: 1,2065 g/cm³ (51 °C) 闪点: >112 °C 存储条件: Ambient |
MDL编号:
MFCD00003327 CAS号: 673-22-3 EINECS: 211-604-0 |
产品技术规格
Appearance | White to beige to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 40 to 43 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...