Volumetric flasks, class B, red markings
供应商: KARTELL
1430 1431 1432 1433 1434 1435
KART1430EA 0 CNY
KART1430 KART1431 KART1432 KART1433 KART1434 KART1435
Volumetric flasks, class B, red markings
烧瓶 容量瓶
PP, translucent, with screw cap.
  • Autoclavable
  • Chemically non absorbant
  • No wetting
  • Suitable for foodstuff
These flasks create no meniscus so final readings are easy. Calibrated gravimetrically at 20 °C. Translucency of PP enhanced when in contact with liquid. Thick walls eliminate distortion.Thin neck make final reading precise.