Torque wrench fittings for Tefzel closures, Nalgene®
供应商: Thermo Fisher Scientific
2195-2028 2195-2020 2195-2024 2195-2033 2195-2038 2195-2043 2195-2048 2195-2053
NALG2195-2028EA 1825 CNY
NALG2195-2028 NALG2195-2020 NALG2195-2024 NALG2195-2033 NALG2195-2038 NALG2195-2043 NALG2195-2048 NALG2195-2053
Torque wrench fittings for Tefzel closures, Nalgene®
扳手
Durable epoxy resin construction, specifically designed for Nalgene ETFE closures.
Proper torquing of Nalgene closures is critical to leakproof sealing.